missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Thienyl)-L-alanine, 95%

Brand:  Alfa Aesar™ H63316.MD

Code : NEW

Additional Details : CAS Number : 22951-96-8 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15499948

  • £36.10 / 250mg


Estimated Shipment date
from Supplier 27-04-2018
Add to basket

Description

Description

Applications
3-(2-Thienyl)-L-alanine can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

3-(2-Thienyl)-L-alanine
171.21
82872
22951-96-8
MFCD00064335
H-3-(2-thienyl)-Ala-OH; (S)-2-Amino-3-(2-thienyl)propionic acid

Specifications

3-(2-Thienyl)-L-alanine
Solid
171.21
82872
Off-white
250mg
22951-96-8
95%
MFCD00064335
H-3-(2-thienyl)-Ala-OH; (S)-2-Amino-3-(2-thienyl)propionic acid
255° to 263°C (dec.)
Videos
SDS