missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Thienyl)-D-alanine, 95%, Thermo Scientific™

Catalog Number 15460499
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
62561-76-6
C7H9NO2S
171.214
MFCD00077166
WTOFYLAWDLQMBZ-ZCFIWIBFSA-N
3-2-thienyl-d-alanine, d-2-thienylalanine, r-2-amino-3-thiophen-2-yl propanoic acid, beta-2-thienyl-d-alanine, h-d-thi-oh, 2r-2-amino-3-thiophen-2-ylpropanoic acid, 2r-2-amino-3-thiophen-2-yl propanoic acid, 2r-2-azanyl-3-thiophen-2-yl-propanoic acid, pubchem18712
7111750
CHEBI:77819
(2R)-2-amino-3-thiophen-2-ylpropanoic acid
C1=CSC(=C1)CC(C(=O)O)N
This item is not returnable. View return policy
62561-76-6
C7H9NO2S
171.214
MFCD00077166
WTOFYLAWDLQMBZ-ZCFIWIBFSA-N
3-2-thienyl-d-alanine, d-2-thienylalanine, r-2-amino-3-thiophen-2-yl propanoic acid, beta-2-thienyl-d-alanine, h-d-thi-oh, 2r-2-amino-3-thiophen-2-ylpropanoic acid, 2r-2-amino-3-thiophen-2-yl propanoic acid, 2r-2-azanyl-3-thiophen-2-yl-propanoic acid, pubchem18712
7111750
CHEBI:77819
(2R)-2-amino-3-thiophen-2-ylpropanoic acid
C1=CSC(=C1)CC(C(=O)O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 171.21
Percent Purity 95%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material 3-(2-Thienyl)-D-alanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'