missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-(2-Thienyl)acrylic acid, predominantly trans, 98+%
Click to view available options
Quantity:
10 g
50 g
Specifications
Melting Point | 145°C to 148°C |
Quantity | 10 g |
Beilstein | 112053 |
Formula Weight | 154.19 |
Percent Purity | ≥98% |
Chemical Name or Material | 3- (2-Thienyl)acrylic acid, predominantly trans |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'