missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Thiazolidinyl)phenol, 97%

Brand:  Alfa Aesar™ H62119.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448698

  • £79.71 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(2-Thiazolidinyl)phenol
181.25
2-(3-Hydroxyphenyl)thiazolidine
C9H11NOS
MFCD16685178

Specifications

3-(2-Thiazolidinyl)phenol
97%
181.25
2-(3-Hydroxyphenyl)thiazolidine
1g
C9H11NOS
MFCD16685178
Videos
SDS