missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Thiazolidinyl)phenol, 97%

Brand:  Alfa Aesar™ H62119.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458698

  • £398.56 / 5g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(2-Thiazolidinyl)phenol
181.25
2-(3-Hydroxyphenyl)thiazolidine
C9H11NOS
MFCD16685178

Specifications

3-(2-Thiazolidinyl)phenol
97%
181.25
2-(3-Hydroxyphenyl)thiazolidine
5g
C9H11NOS
MFCD16685178
Videos
SDS