Learn More
Sartorius™ 3.2 Standard Flexboy™ with TPE tubes
Brand:  Sartorius™ FFB110911
Code : NEW
Additional Details : Weight : 21.00000kg
Product Code. 15938101
  • £5294.70 / Pack of 20
Estimated Shipment: 09-05-2024
to see stock.
Specifications
10 L
Bags are designed for the preparation, storage and transport of biopharmaceutical solutions, intermediates and final bulk products
Gamma Irradiated
ISO11137
3 Ports