missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Pyridyl)-D-alanine, 95%, Thermo Scientific™

Catalog Number 15449918
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
37535-52-7
C8H10N2O2
166.18
MFCD00237245
PDRJLZDUOULRHE-SSDOTTSWSA-N
3-2-pyridyl-d-alanine, r-2-amino-3-pyridin-2-yl propanoic acid, d-2-pyridylalanine, 2r-2-amino-3-pyridin-2-yl propanoic acid, h-d-2-pal-oh, 2'-pyridyl-d-ala, 3-2-pyridyl-d-ala-oh, d-3-2-pyridyl-alanine, r-2-pyridylalanine, 2-pyridyl-d-ala
7019768
(2R)-2-amino-3-pyridin-2-ylpropanoic acid
C1=CC=NC(=C1)CC(C(=O)O)N
This item is not returnable. View return policy
37535-52-7
C8H10N2O2
166.18
MFCD00237245
PDRJLZDUOULRHE-SSDOTTSWSA-N
3-2-pyridyl-d-alanine, r-2-amino-3-pyridin-2-yl propanoic acid, d-2-pyridylalanine, 2r-2-amino-3-pyridin-2-yl propanoic acid, h-d-2-pal-oh, 2'-pyridyl-d-ala, 3-2-pyridyl-d-ala-oh, d-3-2-pyridyl-alanine, r-2-pyridylalanine, 2-pyridyl-d-ala
7019768
(2R)-2-amino-3-pyridin-2-ylpropanoic acid
C1=CC=NC(=C1)CC(C(=O)O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Beilstein 6478361
Formula Weight 166.18
Percent Purity 95%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material 3-(2-Pyridyl)-D-alanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'