missing translation for 'onlineSavingsMsg'
Learn More

3-[2-Oxo-3-(trifluoromethyl)-1,2-dihydropyridin-1-yl]propanenitrile, 97%, Maybridge™

£9.04 - £61.28

Chemical Identifiers

CAS 175277-60-8
Molecular Formula C9H7F3N2O
Molecular Weight (g/mol) 216.163
MDL Number MFCD00204216
InChI Key CQBBWSPMEJVWQE-UHFFFAOYSA-N
Synonym 3-2-oxo-3-trifluoromethyl pyridin-1 2h-yl propanenitrile, 1-2-cyanoethyl-3-trifluoromethyl-2 1h-pyridone, 3-2-oxo-3-trifluoromethyl-1,2-dihydropyridin-1-yl propanenitrile, 3-2-oxo-3-trifluoromethyl pyridin-1-yl propanenitrile, 1-2-cyanoethyl-3-trifluoromethyl pyrid-2 1h-one, 1-2-cyanoethyl-3-trifluoromethyl pyrid-2-1h-one, 1-2-cyanoethyl-3-trifluoromethyl pyridin-2 1h-one, 1-2-cyanoethyl-3-trifluoromethyl-pyrid-2-1h-one, 1-2-cyanoethyl-3-trifluoromethyl-2 1h-pyridon, 3-2-oxo-3-trifluoromethyl hydropyridyl propanenitrile
PubChem CID 2736731
IUPAC Name 3-[2-oxo-3-(trifluoromethyl)pyridin-1-yl]propanenitrile
SMILES C1=CN(C(=O)C(=C1)C(F)(F)F)CCC#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10611893
SDS View Documents
Maybridge
SPB06060DA
1g
£9.04
1g
View alternative products
Save Up to 
»
Estimated Shipment: 21-07-2021
Please to purchase this item. Need a web account? Register with us today!
10019571
SDS View Documents
Maybridge
SPB06060EA
10g
£61.28
10g
View alternative products
Save Up to 
»
Estimated Shipment: 21-07-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

175277-60-8
216.163
CQBBWSPMEJVWQE-UHFFFAOYSA-N
2736731
C1=CN(C(=O)C(=C1)C(F)(F)F)CCC#N
C9H7F3N2O
MFCD00204216
3-2-oxo-3-trifluoromethyl pyridin-1 2h-yl propanenitrile, 1-2-cyanoethyl-3-trifluoromethyl-2 1h-pyridone, 3-2-oxo-3-trifluoromethyl-1,2-dihydropyridin-1-yl propanenitrile, 3-2-oxo-3-trifluoromethyl pyridin-1-yl propanenitrile, 1-2-cyanoethyl-3-trifluoromethyl pyrid-2 1h-one, 1-2-cyanoethyl-3-trifluoromethyl pyrid-2-1h-one, 1-2-cyanoethyl-3-trifluoromethyl pyridin-2 1h-one, 1-2-cyanoethyl-3-trifluoromethyl-pyrid-2-1h-one, 1-2-cyanoethyl-3-trifluoromethyl-2 1h-pyridon, 3-2-oxo-3-trifluoromethyl hydropyridyl propanenitrile
3-[2-oxo-3-(trifluoromethyl)pyridin-1-yl]propanenitrile

Specifications

3-[2-Oxo-3-(trifluoromethyl)-1, 2-dihydropyridin-1-yl]propanenitrile
175277-60-8
97%
MFCD00204216
CQBBWSPMEJVWQE-UHFFFAOYSA-N
3-[2-oxo-3-(trifluoromethyl)pyridin-1-yl]propanenitrile
2736731
97%
1g
97.0
C9H7F3N2O
3-2-oxo-3-trifluoromethyl pyridin-1 2h-yl propanenitrile, 1-2-cyanoethyl-3-trifluoromethyl-2 1h-pyridone, 3-2-oxo-3-trifluoromethyl-1,2-dihydropyridin-1-yl propanenitrile, 3-2-oxo-3-trifluoromethyl pyridin-1-yl propanenitrile, 1-2-cyanoethyl-3-trifluoromethyl pyrid-2 1h-one, 1-2-cyanoethyl-3-trifluoromethyl pyrid-2-1h-one, 1-2-cyanoethyl-3-trifluoromethyl pyridin-2 1h-one, 1-2-cyanoethyl-3-trifluoromethyl-pyrid-2-1h-one, 1-2-cyanoethyl-3-trifluoromethyl-2 1h-pyridon, 3-2-oxo-3-trifluoromethyl hydropyridyl propanenitrile
C1=CN(C(=O)C(=C1)C(F)(F)F)CCC#N
216.163
216.16
SDS