missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Naphthyl)-L-alanine, 98%, ACROS Organics™

Brand:  Acros Organics 333900050

Code : 29

Additional Details : CAS Number : 58438-03-2 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10606734

  • £999.30 / 5g


Estimated Shipment
09-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(2-Naphthyl)-L-alanine
97.5
≥97.5%
C13H13NO2
MFCD00066087
58438-03-2
100.0
98%
215.25

Specifications

3-(2-Naphthyl)-L-alanine
97.5
Crystalline Powder
98%
C13H13NO2
MFCD00066087
Off-white
58438-03-2
100.0
≥97.5%
Authentic
215.25
Glass bottle
5g
Videos
SDS