missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Naphthyl)-L-alanine, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Packaging:
Glass Bottle
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | White |
Assay Percent Range | 98% |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Formula Weight | 215.25 |
Percent Purity | ≥97.5% |
Quantity | 250 mg |
Physical Form | Crystalline Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'