missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Naphthyl)-D-alanine, 95%

Brand:  Alfa Aesar™ H63532.06

Code : NEW

Additional Details : CAS Number : 76985-09-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15450269

  • £172.80 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(2-Naphthyl)-D-alanine
215.25
4352384
76985-09-6
MFCD00038546
H-D-2-Nal-OH

Specifications

3-(2-Naphthyl)-D-alanine
Solid
215.25
4352384
Off-white
76985-09-6
95%
MFCD00038546
H-D-2-Nal-OH
5g
Videos
SDS