missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Morpholinoethoxy)phenyl]methanol, 97%, Maybridge™

POA

Chemical Identifiers

CAS 857284-07-2
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD07772846
InChI Key DZHYOLUKMGNRTO-UHFFFAOYSA-N
Synonym 3-2-morpholinoethoxy phenyl methanol, 3-2-morpholin-4-yl ethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methan-1-ol
PubChem CID 7164622
IUPAC Name [3-(2-morpholin-4-ylethoxy)phenyl]methanol
SMILES C1COCCN1CCOC2=CC=CC(=C2)CO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10249752
SDS View Documents
Maybridge
CC42009CB
250mg
POA
250mg
View alternative products
Save Up to 
»
N/A Please to purchase this item. Need a web account? Register with us today!
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
10015712
SDS View Documents
Maybridge
CC42009DA
1g
POA
1g
View alternative products
Save Up to 
»
N/A Please to purchase this item. Need a web account? Register with us today!
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
Description

Description

Specifications

Chemical Identifiers

857284-07-2
237.299
DZHYOLUKMGNRTO-UHFFFAOYSA-N
7164622
C1COCCN1CCOC2=CC=CC(=C2)CO
C13H19NO3
MFCD07772846
3-2-morpholinoethoxy phenyl methanol, 3-2-morpholin-4-yl ethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methan-1-ol
[3-(2-morpholin-4-ylethoxy)phenyl]methanol

Specifications

[3-(2-Morpholinoethoxy)phenyl]methanol
857284-07-2
100.0
C13H19NO3
3-2-morpholinoethoxy phenyl methanol, 3-2-morpholin-4-yl ethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methanol, 3-2-morpholin-4-ylethoxy phenyl methan-1-ol
C1COCCN1CCOC2=CC=CC(=C2)CO
237.299
237.3
250mg
96.5
97%
MFCD07772846
DZHYOLUKMGNRTO-UHFFFAOYSA-N
[3-(2-morpholin-4-ylethoxy)phenyl]methanol
7164622
97%
SDS
Product Certifications