missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Methyl-thiazol-4-yl)-benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-3110350
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
850375-05-2
C11H9NOS
203.26
MFCD02681946
FQQPVEKQPGDYPD-UHFFFAOYSA-N
7127782
3-(2-methyl-1,3-thiazol-4-yl)benzaldehyde
CC1=NC(=CS1)C1=CC(C=O)=CC=C1
This item is not returnable. View return policy
850375-05-2
C11H9NOS
203.26
MFCD02681946
FQQPVEKQPGDYPD-UHFFFAOYSA-N
7127782
3-(2-methyl-1,3-thiazol-4-yl)benzaldehyde
CC1=NC(=CS1)C1=CC(C=O)=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-2-methyl-thiazol-4-yl-benzaldehyde, 3-2-methyl-1,3-thiazol-4-yl benzaldehyde, 3-2-methylthiazol-4-yl benzaldehyde, 4-3-formylphenyl-2-methyl-1,3-thiazole, 3-2-methyl-4-thiazolyl benzaldehyde, benzaldehyde,3-2-methyl-4-thiazolyl, benzaldehyde, 3-2-methyl-4-thiazolyl
Formula Weight 203.26
Percent Purity 97%
Chemical Name or Material 3-(2-Methyl-thiazol-4-yl)-benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'