missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Methyl-1,3-thiazol-4-yl)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 219.26 |
Percent Purity | 97% |
Chemical Name or Material | 3-(2-Methyl-1, 3-thiazol-4-yl)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'