missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Hydroxyethylsulfamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog Number 15499965
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 913835-57-1
Melting Point 228°C to 230°C
Molecular Formula C8H12BNO5S
MDL Number MFCD08436067
Quantity 250 mg
Synonym 3-n-2-hydroxyethyl sulfamoyl phenyl boronic acid, n-2-hydroxyethyl 3-boronobenzenesulfonamide, 3-n-2-hydroxyethyl sulphamoyl benzeneboronic acid, 3-2-hydroxyethyl sulfamoyl phenylboronic acid, acmc-209r9m, n-2-hydroxyethyl-3-boronobenzenesulfonamide, 3-2-hydroxyethylsulfamoyl benzeneboronic acid, 3-borono-n-2-hydroxyethyl benzenesulphonamide, n-2-hydroxyethyl-3-boronobenzene sulfonamide, 3-2-hydroxyethyl sulfamoyl phenyl boronic acid
InChI Key HSOUGLHWSPWPOM-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)S(=O)(=O)NCCO)(O)O
IUPAC Name [3-(2-hydroxyethylsulfamoyl)phenyl]boronic acid
Molecular Weight (g/mol) 245.056
PubChem CID 44119345
Formula Weight 245.06
Percent Purity 98%
Chemical Name or Material 3-(2-Hydroxyethylsulfamoyl)benzeneboronic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'