Learn More
Thermo Scientific Maybridge [3-(2-Furyl)phenyl]methylamine, 1g, 97%
Brand: Thermo Scientific Maybridge CC46113DA
[3-(2-Furyl)phenyl]methylamine, 1g, 97%
Code : 31
Additional Details : CAS Number : 771573-26-3 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III