missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Furyl)benzonitrile, ≥97%, Thermo Scientific™

Catalog Number p-3175267
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
112598-77-3
C11H7NO
169.183
MFCD04039075
BUZQKXZXWYCNJK-UHFFFAOYSA-N
4163731
3-(furan-2-yl)benzonitrile
C1=CC(=CC(=C1)C2=CC=CO2)C#N
This item is not returnable. View return policy
112598-77-3
C11H7NO
169.183
MFCD04039075
BUZQKXZXWYCNJK-UHFFFAOYSA-N
4163731
3-(furan-2-yl)benzonitrile
C1=CC(=CC(=C1)C2=CC=CO2)C#N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Synonym 3-2-furyl benzonitrile, 3-furan-2-yl benzonitrile, benzonitrile,3-2-furanyl, 3-furan-2-yl-benzonitrile, benzonitrile, 3-2-furanyl, acmc-20mgkx, 3-2-furanyl benzonitrile, 3-2-furyl benzenecarbonitrile, 3-furan-2-yl benzenecarbonitrile
Formula Weight 169.18
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'