missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Furyl)-1H-pyrazol-5-amine, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 149.15 |
Percent Purity | 95% |
Chemical Name or Material | 3-(2-Furyl)-1H-pyrazol-5-amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'