missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 178.19 |
Percent Purity | 97% |
Chemical Name or Material | [3-(2-Furyl)-1-methyl-1H-pyrazol-5-yl]methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'