missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-(2-Fluorophenyl)-1-phenyl-2-thiohydantoin, 95%
Brand: Alfa Aesar™ H64751.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
3-(2-Fluorophenyl)-1-phenyl-2-thiohydantoin | |
286.32 | |
MFCD26967926 | |
3-(2-Fluorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone |
Specifications
3-(2-Fluorophenyl)-1-phenyl-2-thiohydantoin | |
Solid | |
286.32 | |
3-(2-Fluorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone |
1g | |
95% | |
MFCD26967926 |