missing translation for 'onlineSavingsMsg'
Learn More

3-[2-Fluoro-4-(trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™

Catalog Number p-7030981
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
916420-38-7
C10H8F4O2
236.17
MFCD09025355
QMWPFJAQAQWUMS-UHFFFAOYSA-N
3-2-fluoro-4-trifluoromethyl phenyl propionic acid, 3-2-fluoro-4-trifluoromethyl phenyl propanoic acid, 3-2-fluoro-4-trifluoromethyl-phenyl-propionic acid
46737485
3-[2-fluoro-4-(trifluoromethyl)phenyl]propanoic acid
OC(=O)CCC1=C(F)C=C(C=C1)C(F)(F)F
This item is not returnable. View return policy
916420-38-7
C10H8F4O2
236.17
MFCD09025355
QMWPFJAQAQWUMS-UHFFFAOYSA-N
3-2-fluoro-4-trifluoromethyl phenyl propionic acid, 3-2-fluoro-4-trifluoromethyl phenyl propanoic acid, 3-2-fluoro-4-trifluoromethyl-phenyl-propionic acid
46737485
3-[2-fluoro-4-(trifluoromethyl)phenyl]propanoic acid
OC(=O)CCC1=C(F)C=C(C=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 59°C to 61°C
Quantity 250 mg
Formula Weight 236.16
Percent Purity 97%
Chemical Name or Material 3-[2-Fluoro-4-(trifluoromethyl)phenyl]propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'