missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Chlorophenyl)-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63061.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15429538

  • £398.56 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(2-Chlorophenyl)-2-thiohydantoin
226.68
MFCD03070245
3-(2-Chlorophenyl)-2-thioxo-4-imidazolidinone

Specifications

3-(2-Chlorophenyl)-2-thiohydantoin
Solid
226.68
3-(2-Chlorophenyl)-2-thioxo-4-imidazolidinone
5g
95%
MFCD03070245
Videos
SDS