missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Chlorophenyl)-1-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64172.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15441259

  • £259.70 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

3-(2-Chlorophenyl)-1-phenyl-2-thiohydantoin
302.78
MFCD27976895
3-(2-Chlorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone

Specifications

3-(2-Chlorophenyl)-1-phenyl-2-thiohydantoin
Solid
302.78
3-(2-Chlorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone
5g
95%
MFCD27976895
Videos
SDS