missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Chlorophenyl)-1-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64172.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15431259

  • £51.94 / 1g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(2-Chlorophenyl)-1-phenyl-2-thiohydantoin
302.78
MFCD27976895
3-(2-Chlorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone

Specifications

3-(2-Chlorophenyl)-1-phenyl-2-thiohydantoin
Solid
302.78
3-(2-Chlorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone
1g
95%
MFCD27976895
Videos
SDS