missing translation for 'onlineSavingsMsg'
Learn More

3-(2-chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide, 97%, Thermo Scientific™

Catalog Number p-2698966
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
4415-11-6
C11H8ClFN2O2
254.65
MFCD00173954
PPKCMGUTHICRLU-UHFFFAOYSA-N
3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxamide, 3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxamide, maybridge1_006010, 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4carboxamide, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxamide, 3-6-chloro-2-fluorophenyl-5-methylisoxazole-4-carboxamide, 4-isoxazolecarboxamide,3-2-chloro-6-fluorophenyl-5-methyl
2736583
This item is not returnable. View return policy
4415-11-6
C11H8ClFN2O2
254.65
MFCD00173954
PPKCMGUTHICRLU-UHFFFAOYSA-N
3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxamide, 3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxamide, maybridge1_006010, 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4carboxamide, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxamide, 3-6-chloro-2-fluorophenyl-5-methylisoxazole-4-carboxamide, 4-isoxazolecarboxamide,3-2-chloro-6-fluorophenyl-5-methyl
2736583
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 254.65
Percent Purity 97%
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'