missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2-Bromophenyl)-1-propanol, 98%

Brand:  Alfa Aesar™ H63884.06

Code : NEW

Additional Details : CAS Number : 52221-92-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15420839

  • £289.90 / 5g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(2-Bromophenyl)-1-propanol
52221-92-8
215.09
MFCD09028685

Specifications

3-(2-Bromophenyl)-1-propanol
5g
98%
215.09
52221-92-8
Liquid
Colorless
MFCD09028685
Videos
SDS