missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Bromoacetyl)benzonitrile, 95%, Thermo Scientific™

Catalog Number 10722513
Click to view available options
Quantity:
1 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
50916-55-7
C9H6BrNO
224.06
MFCD00833267
XWCGNFLHRINYCE-UHFFFAOYSA-N
3-2-bromoacetyl benzonitrile, 3-cyanophenacyl bromide, 3-cyanophenacylbromide, 2-bromo-3'-cyanoacetophenone, 3-2-bromoacetyl benzenecarbonitrile, 3-bromoacetyl benzonitrile, 3-2-bromo-acetyl-benzonitrile, benzonitrile, 3-bromoacetyl, 3-bromoacetyl-benzonitrile
2735867
3-(2-bromoacetyl)benzonitrile
BrCC(=O)C1=CC=CC(=C1)C#N
This item is not returnable. View return policy
50916-55-7
C9H6BrNO
224.06
MFCD00833267
XWCGNFLHRINYCE-UHFFFAOYSA-N
3-2-bromoacetyl benzonitrile, 3-cyanophenacyl bromide, 3-cyanophenacylbromide, 2-bromo-3'-cyanoacetophenone, 3-2-bromoacetyl benzenecarbonitrile, 3-bromoacetyl benzonitrile, 3-2-bromo-acetyl-benzonitrile, benzonitrile, 3-bromoacetyl, 3-bromoacetyl-benzonitrile
2735867
3-(2-bromoacetyl)benzonitrile
BrCC(=O)C1=CC=CC(=C1)C#N
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula BrCH2C(O)C6H4CN
Quantity 1 g
Formula Weight 224.06
Percent Purity 95%
Chemical Name or Material 3-(2-Bromoacetyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'