missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 254.08 |
Percent Purity | 97% |
Chemical Name or Material | 3-(2-Bromo-5-methoxyphenyl)-3-oxopropanenitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'