missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2,6-Difluorophenyl)-2-thioxo-4-thiazolidinone, 95%

Brand:  Alfa Aesar™ H63610.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428406

  • £460.04 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(2,6-Difluorophenyl)-2-thioxo-4-thiazolidinone
245.26
MFCD27975660
3-(2,6-Difluorophenyl)rhodanine

Specifications

3-(2,6-Difluorophenyl)-2-thioxo-4-thiazolidinone
Solid
245.26
3-(2,6-Difluorophenyl)rhodanine
5g
95%
MFCD27975660
Videos
SDS