missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(2,5-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

Brand:  Alfa Aesar™ H56125.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15432496

  • £358.49 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(2,5-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid
287.09
MFCD27665219

Specifications

3-(2,5-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid
Solid
287.09
1g
97%
MFCD27665219
Videos
SDS