missing translation for 'onlineSavingsMsg'
Learn More

3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech., Thermo Scientific Chemicals

Catalog Number p-3756793
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
35141-30-1
C10H27N3O3Si
265.43
MFCD00069173
NHBRUUFBSBSTHM-UHFFFAOYSA-N
n1-2-aminoethyl-n2-3-trimethoxysilyl propyl ethane-1,2-diamine, 3-trimethoxysilylpropyl diethylenetriamine, 3-2-2-aminoethylamino ethylamino propyl-trimethoxysilane, 3-trimethoxysilyl propyl diethylenetriamine, n1-3-trimethoxysilylpropyl diethylenetriamine, trimethoxysilylpropyldiethylenetriamine, 1,2-ethanediamine, n-2-aminoethyl-n'-3-trimethoxysilyl propyl, diethylenetriamino propyltrimethoxysilane, 3-2-2-aminoethylamino ethylamino propyltrimethoxysilane, n-2-aminoethyl-n'-3-trimethoxysilyl propyl ethylenediamine
118771
N'-[2-(3-trimethoxysilylpropylamino)ethyl]ethane-1,2-diamine
CO[Si](CCCNCCNCCN)(OC)OC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
35141-30-1
C10H27N3O3Si
265.43
MFCD00069173
NHBRUUFBSBSTHM-UHFFFAOYSA-N
n1-2-aminoethyl-n2-3-trimethoxysilyl propyl ethane-1,2-diamine, 3-trimethoxysilylpropyl diethylenetriamine, 3-2-2-aminoethylamino ethylamino propyl-trimethoxysilane, 3-trimethoxysilyl propyl diethylenetriamine, n1-3-trimethoxysilylpropyl diethylenetriamine, trimethoxysilylpropyldiethylenetriamine, 1,2-ethanediamine, n-2-aminoethyl-n'-3-trimethoxysilyl propyl, diethylenetriamino propyltrimethoxysilane, 3-2-2-aminoethylamino ethylamino propyltrimethoxysilane, n-2-aminoethyl-n'-3-trimethoxysilyl propyl ethylenediamine
118771
N'-[2-(3-trimethoxysilylpropylamino)ethyl]ethane-1,2-diamine
CO[Si](CCCNCCNCCN)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.0310g/mL
Boiling Point 114.0°C to 118.0°C (2.0 mmHg)
Flash Point 137°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (H3CO)3Si(CH2)3NH(CH2)2NH(CH2)2NH2
Refractive Index 1.4570 to 1.4610
Quantity 25 mL
Specific Gravity 1.031
Solubility Information Solubility in water: immiscible
Formula Weight 265.43
Percent Purity 14 to 18% (amine (as NH2))
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-¢2-(2-Aminoethylamino)ethylamino!propyl-trimethoxysilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'