missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-(1H-Tetrazol-5-yl)benzyl alcohol, 96%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 208°C to 210°C |
Quantity | 250 mg |
Formula Weight | 176.18 |
Percent Purity | 96% |
Chemical Name or Material | 3-(1H-Tetrazol-5-yl)benzyl alcohol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'