missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)propanenitrile, 97%, Maybridge

Brand:  Maybridge CD01949DA

Code : 31

Additional Details : CAS Number : 43036-06-2 Weight : 0.05100kg

Quantity 1g

 View more versions of this product

Product Code. 10673625

  • £7.21 / 1g


Estimated Shipment date
from Supplier 23-01-2018
Add to basket

Specifications

Chemical Identifiers

3-(1H-Pyrrol-1-yl)propanenitrile
C7H8N2
1-Pyrrolepropionitrile
97%
120.15

Specifications

3-(1H-Pyrrol-1-yl)propanenitrile
43036-06-2
C7H8N2
1-Pyrrolepropionitrile
1g
97%
120.15
Videos
SDS