missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-2698869
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
61471-45-2
C11H8NO2
186.19
MFCD02656610
PODFNQCZFHLJPH-UHFFFAOYSA-M
736537
[O-]C(=O)C1=CC=CC(=C1)N1C=CC=C1
This item is not returnable. View return policy
61471-45-2
C11H8NO2
186.19
MFCD02656610
PODFNQCZFHLJPH-UHFFFAOYSA-M
736537
[O-]C(=O)C1=CC=CC(=C1)N1C=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 3-1h-pyrrol-1-yl benzoic acid, 3-pyrrol-1-yl-benzoic acid, benzoic acid, 3-1h-pyrrol-1-yl, n-3-carboxy phenylpyrrole, 3-1-pyrrolyl benzoic acid, 3-pyrrol-1-yl benzoic acid, 3-pyrrolylbenzoic acid, 1-3-carboxyphenyl-1h-pyrrole, benzoicacid,3-1h-pyrrol-1-yl, benzoicacid, 3-1h-pyrrol-1-yl
IUPAC Name 3-(1H-pyrrol-1-yl)benzoate
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 3-(1H-Pyrrol-1-yl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'