missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrrol-1-yl)benzoic acid, 97%, Maybridge

Brand:  Maybridge CC21901EA

UNSPSC : 12350000

Code : 31

Additional Details : CAS Number : 61471-45-2 Weight : 0.06000kg

Quantity 10g

 View more versions of this product

Product Code. 10265704

  • £302.00 / 10g


Estimated Shipment date
from Supplier 29-06-2017
Add to basket


Specifications Description & Specifications

Chemical Identifiers

3-(1H-Pyrrol-1-yl)benzoic acid
96.5
97%
C11H9NO2
MFCD02656610
61471-45-2
100.0
97%
187.2

Specifications

187.2
MFCD02656610
96.5
61471-45-2
C11H9NO2
10g
97%
3-(1H-Pyrrol-1-yl)benzoic acid
97%
100.0
Documentation

Technical Documents