missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Pyrazol-3-yl)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-4510472
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
850375-11-0
C10H8N2O2
188.19
MFCD03407956
RXZRBZWATRFHCS-UHFFFAOYSA-N
3-1h-pyrazol-5-yl benzoic acid, 3-1h-pyrazol-3-yl benzoic acid, 3-2h-pyrazol-3-yl benzoic acid, benzoic acid, 3-1h-pyrazol-3-yl, 5-3-carboxyphenyl-1h-pyrazole, pubchem22748, buttpark 44\01-87, d0tx9p, 3-pyrazol-3-ylbenzoic acid, 3-pyrazol-5-ylbenzoic acid
2795542
3-(1H-pyrazol-5-yl)benzoic acid
OC(=O)C1=CC=CC(=C1)C1=CC=NN1
This item is not returnable. View return policy
850375-11-0
C10H8N2O2
188.19
MFCD03407956
RXZRBZWATRFHCS-UHFFFAOYSA-N
3-1h-pyrazol-5-yl benzoic acid, 3-1h-pyrazol-3-yl benzoic acid, 3-2h-pyrazol-3-yl benzoic acid, benzoic acid, 3-1h-pyrazol-3-yl, 5-3-carboxyphenyl-1h-pyrazole, pubchem22748, buttpark 44\01-87, d0tx9p, 3-pyrazol-3-ylbenzoic acid, 3-pyrazol-5-ylbenzoic acid
2795542
3-(1H-pyrazol-5-yl)benzoic acid
OC(=O)C1=CC=CC(=C1)C1=CC=NN1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 3-(1H-Pyrazol-3-yl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'