missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Imidazol-1-ylmethyl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog Number p-3175359
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
102432-05-3
C11H10N2O
186.214
MFCD08271917
JMKQKKQFLYPQDD-UHFFFAOYSA-N
10397537
3-(imidazol-1-ylmethyl)benzaldehyde
C1=CC(=CC(=C1)CN2C=CN=C2)C=O
This item is not returnable. View return policy
102432-05-3
C11H10N2O
186.214
MFCD08271917
JMKQKKQFLYPQDD-UHFFFAOYSA-N
10397537
3-(imidazol-1-ylmethyl)benzaldehyde
C1=CC(=CC(=C1)CN2C=CN=C2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 3-1h-imidazol-1-ylmethyl benzaldehyde, benzaldehyde,3-1h-imidazol-1-ylmethyl, 3-imidazol-1-ylmethyl benzaldehyde, 3-1h-imidazol-1-yl methyl benzaldehyde, benzaldehyde, 3-1h-imidazol-1-ylmethyl, acmc-1c5ro, 3-imidazolylmethyl benzaldehyde
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 3-(1H-imidazol-1-ylmethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'