missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Maybridge 3-(1H-Imidazol-1-yl)benzoic acid, 1g, 97%

Brand:  Thermo Scientific Maybridge CC32601DA

3-(1H-Imidazol-1-yl)benzoic acid, 1g, 97%

Code : 31

Additional Details : Weight : 0.05100kg

Product Code. 10507493

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.