Learn More
Thermo Scientific Maybridge 3-(1H-Imidazol-1-yl)benzoic acid, 5g, 97%
Brand: Thermo Scientific Maybridge CC32601DE
3-(1H-Imidazol-1-yl)benzoic acid, 5g, 97%
Code : 31
Additional Details : CAS Number : 108035-47-8 Weight : 0.05500kg
Brand: Thermo Scientific Maybridge CC32601DE
3-(1H-Imidazol-1-yl)benzoic acid, 5g, 97%
Code : 31
Additional Details : CAS Number : 108035-47-8 Weight : 0.05500kg