missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-1,2,4-triazol-1-ylmethyl)aniline, ≥97%, Thermo Scientific™

Catalog Number p-3176197
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
127988-22-1
C9H10N4
174.207
MFCD08060528
LFINNEVQJQXPHN-UHFFFAOYSA-N
3-1h-1,2,4-triazol-1-ylmethyl aniline, 3-1,2,4-triazol-1-ylmethyl aniline, 3-1,2,4-triazol-1-yl methyl aniline, 3-1,2,4 triazol-1-ylmethyl-phenylamine, 3-1h-1,2,4-triazol-1-yl methyl aniline, benzenamine,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazolylmethyl phenylamine, acmc-20e0ac, 1-3-aminobenzyl-1h-1,2,4-triazole
6482004
3-(1,2,4-triazol-1-ylmethyl)aniline
C1=CC(=CC(=C1)N)CN2C=NC=N2
This item is not returnable. View return policy
127988-22-1
C9H10N4
174.207
MFCD08060528
LFINNEVQJQXPHN-UHFFFAOYSA-N
3-1h-1,2,4-triazol-1-ylmethyl aniline, 3-1,2,4-triazol-1-ylmethyl aniline, 3-1,2,4-triazol-1-yl methyl aniline, 3-1,2,4 triazol-1-ylmethyl-phenylamine, 3-1h-1,2,4-triazol-1-yl methyl aniline, benzenamine,3-1h-1,2,4-triazol-1-ylmethyl, 3-1,2,4-triazolylmethyl phenylamine, acmc-20e0ac, 1-3-aminobenzyl-1h-1,2,4-triazole
6482004
3-(1,2,4-triazol-1-ylmethyl)aniline
C1=CC(=CC(=C1)N)CN2C=NC=N2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 174.21
Percent Purity 97%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-ylmethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'