missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-1,2,4-Triazol-1-yl)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-3176194
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
167626-64-4
C9H7N3O2
189.174
MFCD08271895
SZKWCOCFEIVCAB-UHFFFAOYSA-N
22224944
3-(1,2,4-triazol-1-yl)benzoic acid
C1=CC(=CC(=C1)N2C=NC=N2)C(=O)O
This item is not returnable. View return policy
167626-64-4
C9H7N3O2
189.174
MFCD08271895
SZKWCOCFEIVCAB-UHFFFAOYSA-N
22224944
3-(1,2,4-triazol-1-yl)benzoic acid
C1=CC(=CC(=C1)N2C=NC=N2)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-1h-1,2,4-triazol-1-yl benzoic acid, 3-1,2,4-triazol-1-yl benzoic acid, 3-1,2,4 triazol-1-yl-benzoic acid, benzoic acid, 3-1h-1,2,4-triazol-1-yl, pubchem19567, 3-1,2,4-triazolyl benzoic acid, 1-3-carboxyphenyl-1h-1,2,4-triazole, benzoic acid,3-1h-1,2,4-triazol-1-yl
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 3-(1H-1, 2, 4-Triazol-1-yl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'