missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1h-1,2,4-Triazol-1-yl)aniline, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 3-1h-1,2,4-triazol-1-yl aniline, 3-1,2,4-triazol-1-yl aniline, benzenamine,3-1h-1,2,4-triazol-1-yl, benzenamine, 3-1h-1,2,4-triazol-1-yl, 1-3-aminophenyl-1h-1,2,4-triazole, 3-1,2,4-triazolyl phenylamine, 3-1,2,4 triazol-1-yl-phenylamine, 3-1,2,4 triazol-1-yl phenylamine |
Formula Weight | 160.18 |
Percent Purity | 90% |
Chemical Name or Material | 3-(1H-1, 2, 4-Triazol-1-yl)aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'