missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Pyrrolidinylmethyl)indole, 95%

Brand:  Alfa Aesar™ H63840.03

Code : NEW

Additional Details : CAS Number : 5379-94-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410759

  • £133.70 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Description

Description

Applications
Reatant in the synthesis 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, bases.
Specifications

Chemical Identifiers

3-(1-Pyrrolidinylmethyl)indole
200.28
477183
5379-94-2
MFCD00450050

Specifications

3-(1-Pyrrolidinylmethyl)indole
5379-94-2
95%
MFCD00450050
White to off-white
1g
Solid
200.28
477183
Videos
SDS