missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Naphthyl)-L-alanine, 95%

Brand:  Alfa Aesar™ H63586.06

Code : NEW

Additional Details : CAS Number : 55516-54-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15410359

  • £128.00 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

3-(1-Naphthyl)-L-alanine
215.25
4352833
55516-54-6
MFCD00066086
H-1-Nal-OH; (S)-2-Amino-3-(1-naphthyl)propionic acid

Specifications

3-(1-Naphthyl)-L-alanine
Solid
215.25
4352833
5g
55516-54-6
95%
MFCD00066086
H-1-Nal-OH; (S)-2-Amino-3-(1-naphthyl)propionic acid
Videos
SDS