missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Methyl-4-piperidinyl)indole, 97%

Brand:  Alfa Aesar™ H63020.03

Code : NEW

Additional Details : CAS Number : 17403-07-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439458

  • £129.60 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Methyl-4-piperidinyl)indole
214.31
1573327
17403-07-5
MFCD07776765

Specifications

3-(1-Methyl-4-piperidinyl)indole
17403-07-5
97%
MFCD07776765
Off-white to yellow
1g
Solid
214.31
1573327
Videos
SDS