3-(1-Methyl-1H-pyrazol-3-yl)benzylamine, 97%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC60613DA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 910037-10-4 Weight: 0.05100kg Transport: UN number : 2735 Chem class : 1b Pack group : III


Packaging Amber Glass Bottle
Quantity 1g

Product Code. 10263293

Quantity Price
1 £ 91.0 / 1g
Estimated Shipment date
from Supplier 01-06-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-(1-Methyl-1H-pyrazol-3-yl)benzylamine
CAS 910037-10-4
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C11H13N3
Formula Weight 187.24
MDL Number MFCD09025905

Specification

Formula Weight 187.24
Quantity 1g
CAS 910037-10-4
Assay 90%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 90%
CAS Min % 89.5
CAS Max % 100.0
Packaging Amber Glass Bottle
Chemical Name or Material 3-(1-Methyl-1H-pyrazol-3-yl)benzylamine
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C11H13N3
MDL Number MFCD09025905
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.