missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1-methyl-1H-pyrazol-3-yl)aniline, 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 173.22 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1-Methyl-1H-pyrazol-3-yl)aniline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'