missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Methyl-1,2,3,6-tetrahydro-4-pyridyl)indole, 97%

Brand:  Alfa Aesar™ H63479.06

Code : NEW

Additional Details : CAS Number : 17403-03-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428336

  • £239.56 / 5g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Methyl-1,2,3,6-tetrahydro-4-pyridyl)indole
212.29
912061
17403-03-1
MFCD11840338

Specifications

3-(1-Methyl-1,2,3,6-tetrahydro-4-pyridyl)indole
17403-03-1
97%
MFCD11840338
5g
Solid
212.29
912061
Videos
SDS