missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Methyl-1,2,3,6-tetrahydro-4-pyridyl)-6-nitroindole, 97%

Brand:  Alfa Aesar™ H63467.03

Code : NEW

Additional Details : CAS Number : 200714-15-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498316

  • £95.61 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Methyl-1,2,3,6-tetrahydro-4-pyridyl)-6-nitroindole
200714-15-4
257.29
MFCD23380601

Specifications

3-(1-Methyl-1,2,3,6-tetrahydro-4-pyridyl)-6-nitroindole
1g
Solid
257.29
Yellow
200714-15-4
97%
MFCD23380601
Videos
SDS