missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Methyl-1,2,3,6-tetrahydro-4-pyridyl)-5-nitroindole, 97%

Brand:  Alfa Aesar™ H63777.03

Code : NEW

Additional Details : CAS Number : 106516-27-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498506

  • £59.78 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(1-Methyl-1,2,3,6-tetrahydro-4-pyridyl)-5-nitroindole
257.29
9271462
106516-27-2
MFCD11042722

Specifications

3-(1-Methyl-1,2,3,6-tetrahydro-4-pyridyl)-5-nitroindole
106516-27-2
97%
MFCD11042722
1g
Solid
257.29
9271462
Videos
SDS